KEY465194881 Key Organics / BIONET

2-({4-[(E)-2-phenylethenyl]pyrimidin-2-yl}sulfanyl)acetonitrile

  • MDL number MFCD00244241
  • Cas Number : 338407-73-1
  • Molecular Weight : 253.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70952

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-088_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart