KEY465194892 Key Organics / BIONET

5-methyl-2-(pyrazin-2-yl)-1,3-thiazol-4-yl thiophene-2-carboxylate

  • MDL number MFCD00172597
  • Cas Number : 338409-37-3
  • Molecular Weight : 303.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71057

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-415S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart