KEY465194894 Key Organics / BIONET

5-methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl cyclopropanecarboxylate

  • MDL number MFCD01316546
  • Cas Number : 338409-45-3
  • Molecular Weight : 327.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-419S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart