KEY465194902 Key Organics / BIONET

4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}butanehydrazide

  • MDL number MFCD00172610
  • Cas Number : 332361-09-8
  • Molecular Weight : 296.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71125

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-908_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart