KEY465194905 Key Organics / BIONET

4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-N'-(2-cyanoethanimidoyl)butanehydrazide

  • MDL number MFCD00172612
  • Cas Number : 338410-08-5
  • Molecular Weight : 362.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71144

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart