KEY465194908 Key Organics / BIONET

3,5-diethyl 2-(3,5-dimethyl-1H-pyrazol-1-yl)-6-hydroxypyridine-3,5-dicarboxylate

  • MDL number MFCD00202772
  • Cas Number : 321570-99-4
  • Molecular Weight : 333.344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71162

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-950_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart