KEY465194911 Key Organics / BIONET

1-(4-chlorophenyl)-3-[2-(2-nitroethyl)phenyl]urea

  • MDL number MFCD00974278
  • Cas Number : 338410-35-8
  • Molecular Weight : 319.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71177

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart