KEY465194925 Key Organics / BIONET

4-chloro-N-(1,5,6-trimethyl-1H-1,3-benzodiazol-4-yl)benzene-1-sulfonamide

  • MDL number MFCD00231372
  • Cas Number : 338410-91-6
  • Molecular Weight : 349.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71253

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-361S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart