KEY465194927 Key Organics / BIONET

5-methoxy-2-(methylsulfanyl)pyrimidin-4-ol

  • MDL number MFCD00184817
  • Cas Number : 1671-08-5
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methoxy-2-(methylsulfanyl)-4-pyrimidinol
Inchi Code 1S/C6H8N2O2S/c1-10-4-3-7-6(11-2)8-5(4)9/h3H,1-2H3,(H,7,8,9)
Inchi Key PCFMDDLVLHIRNK-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71265

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3J-374S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart