KEY465194928 Key Organics / BIONET

2-(benzylsulfanyl)-5-methoxypyrimidin-4-ol

  • MDL number MFCD00215607
  • Cas Number : 56495-12-6
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71267

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3J-376S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart