KEY465194929 Key Organics / BIONET

5-methoxy-2-({[3-(trifluoromethyl)phenyl]methyl}sulfanyl)pyrimidin-4-ol

  • MDL number MFCD00215608
  • Cas Number : 338411-00-0
  • Molecular Weight : 316.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71268

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-377S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart