KEY465194937 Key Organics / BIONET

3,7-dibenzyl-3,7-diazabicyclo[3.3.1]nonan-9-one

  • MDL number MFCD00795087
  • Cas Number : 59009-70-0
  • Molecular Weight : 320.436

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71365

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3J-901_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart