KEY465194946 Key Organics / BIONET

2-chloro-1-(5-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}thiophen-2-yl)ethan-1-one

  • MDL number MFCD00974279
  • Cas Number : 338411-65-7
  • Molecular Weight : 354.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71415

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart