KEY465194947 Key Organics / BIONET

10-[(E)-2-[(2,4-difluorophenyl)amino]ethenyl]-2,3,7,11-tetraazatricyclo[7.4.0.0^{2,6}]trideca-1(9),3,5,7,10,12-hexaene-5-carbonitrile

  • MDL number MFCD00664264
  • Cas Number : 338411-69-1
  • Molecular Weight : 348.317

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71417

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart