KEY465194950 Key Organics / BIONET

2-(2-cyano-1H-pyrrol-1-yl)-4-methoxypyridine-3-carbonitrile

  • MDL number MFCD00975457
  • Cas Number : 338411-83-9
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71425

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart