KEY465194951 Key Organics / BIONET

4-methoxy-2-{2-[(1E)-(methoxyimino)methyl]-1H-pyrrol-1-yl}pyridine-3-carbonitrile

  • MDL number MFCD00975458
  • Cas Number : 338411-87-3
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71429

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart