KEY465194952 Key Organics / BIONET

ethyl 4-(4-chlorophenoxy)-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD00206745
  • Cas Number : 439097-52-6
  • Molecular Weight : 345.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71430

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart