KEY465194955 Key Organics / BIONET

{1-methyl-5-[(4-methylphenyl)sulfanyl]-3-phenyl-1H-pyrazol-4-yl}methanol

  • MDL number MFCD00244904
  • Cas Number : 318233-99-7
  • Molecular Weight : 310.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71469

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-355S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart