KEY465194961 Key Organics / BIONET

2-(4-chlorophenyl)-1-[(2,4-dichlorophenyl)methanesulfonyl]propan-2-ol

  • MDL number MFCD00244872
  • Cas Number : 338412-42-3
  • Molecular Weight : 393.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71482

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-371S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart