KEY465194964 Key Organics / BIONET

1-benzyl-N-(2,6-dichlorophenyl)-6-hydroxy-2-oxo-1,2-dihydropyrimidine-4-carboxamide

  • MDL number MFCD00244542
  • Cas Number : 338412-53-6
  • Molecular Weight : 390.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71495

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-515S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart