KEY465194966 Key Organics / BIONET

methyl 2-{[(5-chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methyl]sulfanyl}benzoate

  • MDL number MFCD00663962
  • Cas Number : 318234-12-7
  • Molecular Weight : 372.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71539

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-594S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart