KEY465194970 Key Organics / BIONET

(5-chloro-1-methyl-3-phenyl-1H-pyrazol-4-yl)methyl 2,4-dichlorobenzoate

  • MDL number MFCD00244917
  • Cas Number : 318234-21-8
  • Molecular Weight : 395.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71561

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-644S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart