KEY465194971 Key Organics / BIONET

(5E)-4-hydroxy-5-(hydroxyimino)-2,7-dimethyl-5H-pyrano[2,3-b]pyridin-8-ium-8-olate

  • MDL number MFCD01568654
  • Cas Number : 161153-99-7
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart