KEY465194972 Key Organics / BIONET

3-{2-[1-(3-nitropyridin-2-yl)piperidin-4-ylidene]hydrazin-1-yl}-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD00215508
  • Cas Number : 338412-87-6
  • Molecular Weight : 379.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71571

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3K-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart