KEY465194974 Key Organics / BIONET

3-chloro-4-[2,6-dinitro-4-(trifluoromethyl)phenoxy]aniline

  • MDL number MFCD00244316
  • Cas Number : 338412-96-7
  • Molecular Weight : 377.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71580

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart