KEY465194975 Key Organics / BIONET

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]sulfanyl}-5-[(2Z)-2-(methoxyimino)ethyl]-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00244326
  • Cas Number : 338413-08-4
  • Molecular Weight : 392.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71591

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart