KEY465194976 Key Organics / BIONET

N-[3-chloro-4-(pyrimidin-2-yloxy)phenyl]-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD00244334
  • Cas Number : 338413-19-7
  • Molecular Weight : 391.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71599

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart