KEY465194983 Key Organics / BIONET

1-(2,6-dimethylphenyl)-3-[(E)-2-(furan-2-yl)ethenyl]urea

  • MDL number MFCD00794359
  • Cas Number : 478031-02-6
  • Molecular Weight : 256.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71637

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart