KEY465194987 Key Organics / BIONET

1-[(E)-2-(4-chlorophenyl)ethenyl]-3-[(2-methoxyphenyl)methyl]urea

  • MDL number MFCD00794848
  • Cas Number : 338413-52-8
  • Molecular Weight : 316.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71673

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-380S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart