KEY465194991 Key Organics / BIONET

3-benzoyl-4-(4-bromophenyl)-1H-pyrrole

  • MDL number MFCD00794455
  • Cas Number : 478031-23-1
  • Molecular Weight : 326.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71696

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-506S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart