KEY465194993 Key Organics / BIONET

3-(4-chlorobenzoyl)-4-(4-chlorophenyl)-1H-pyrrole

  • MDL number MFCD00794461
  • Cas Number : 170939-22-7
  • Molecular Weight : 316.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-512S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart