KEY465194996 Key Organics / BIONET

3-(3,4-dichlorobenzoyl)-4-(3-fluorophenyl)-1H-pyrrole

  • MDL number MFCD00794469
  • Cas Number : 478031-40-2
  • Molecular Weight : 334.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-528S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart