KEY465195002 Key Organics / BIONET

N-benzyl-6-[(methylsulfanyl)methyl]-2-(pyridin-4-yl)pyrimidin-4-amine

  • MDL number MFCD00794481
  • Cas Number : 478031-50-4
  • Molecular Weight : 322.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71730

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-549S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart