KEY465195003 Key Organics / BIONET

(4-chlorophenyl)[4-(2,6-dichlorophenyl)-1H-pyrrol-3-yl]methanol

  • MDL number MFCD00794766
  • Cas Number : 338413-76-6
  • Molecular Weight : 352.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71739

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3L-567S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart