KEY465195007 Key Organics / BIONET

ethyl (2Z)-3-amino-2-cyanoprop-2-enoate

  • MDL number MFCD01936248
  • Cas Number : 67710-34-3
  • Molecular Weight : 140.142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71788

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-906_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart