KEY465195008 Key Organics / BIONET

5-amino-6-nitroso-7H-[1,3]thiazolo[3,2-a]pyrimidin-7-one

  • MDL number MFCD00794648
  • Cas Number : 174279-85-7
  • Molecular Weight : 196.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71805

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-924_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart