KEY465195014 Key Organics / BIONET

(2E)-3-(4-bromophenyl)-2-[2-(4-chlorophenyl)hydrazin-1-ylidene]-3-oxopropanal

  • MDL number MFCD00975105
  • Cas Number : 338414-04-3
  • Molecular Weight : 365.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71813

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-009_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart