KEY465195021 Key Organics / BIONET

(2E)-3-(4-methoxyphenyl)-3-oxo-2-(2-phenylhydrazin-1-ylidene)propanal

  • MDL number MFCD00975114
  • Cas Number : 60083-35-4
  • Molecular Weight : 282.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71826

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart