KEY465195023 Key Organics / BIONET

(2E,3E)-3-hydrazinylidene-1-(4-methoxyphenyl)-2-{2-[4-(trifluoromethyl)phenyl]hydrazin-1-ylidene}propan-1-one

  • MDL number MFCD00975115
  • Cas Number : 338414-19-0
  • Molecular Weight : 364.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71828

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart