KEY465195024 Key Organics / BIONET

4-(4-chlorophenyl)-5-[(Z)-2-(4-methylphenyl)diazen-1-yl]pyrimidin-2-amine

  • MDL number MFCD00974771
  • Cas Number : 338414-20-3
  • Molecular Weight : 323.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71829

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart