KEY465195026 Key Organics / BIONET

(2E,3E)-2-[2-(3,5-dichlorophenyl)hydrazin-1-ylidene]-3-(N-hydroxyimino)-1-(4-methylphenyl)propan-1-one

  • MDL number MFCD00975117
  • Cas Number : 338414-24-7
  • Molecular Weight : 350.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71833

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart