KEY465195027 Key Organics / BIONET

(4E)-3-(furan-2-yl)-4-[2-(3-methylphenyl)hydrazin-1-ylidene]-4H-pyrazole

  • MDL number MFCD00974670
  • Cas Number : 338414-25-8
  • Molecular Weight : 252.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71834

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart