KEY465195038 Key Organics / BIONET

(3E)-3-{[(4-methylphenyl)methoxy]imino}-1-(2-phenyl-1,3-thiazol-5-yl)propan-1-one

  • MDL number MFCD00974291
  • Cas Number : 338414-40-7
  • Molecular Weight : 350.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-046_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart