KEY465195041 Key Organics / BIONET

(2E)-3-(2,4-dichlorophenyl)-3-oxo-2-(2-phenylhydrazin-1-ylidene)propanal

  • MDL number MFCD00975124
  • Cas Number : 338414-48-5
  • Molecular Weight : 321.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71856

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart