KEY465195044 Key Organics / BIONET

(2E)-3-[(4-fluorophenyl)amino]-1-(2-phenyl-1,3-thiazol-5-yl)prop-2-en-1-one

  • MDL number MFCD00974297
  • Cas Number : 338414-54-3
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71859

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart