KEY465195048 Key Organics / BIONET

(4E)-3-(4-chlorophenyl)-4-[2-(4-chlorophenyl)hydrazin-1-ylidene]-4H-pyrazole

  • MDL number MFCD00974676
  • Cas Number : 338414-63-4
  • Molecular Weight : 317.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71868

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart