KEY465195050 Key Organics / BIONET

(3Z)-3-[(3-nitrophenyl)imino]-1-(prop-2-en-1-yl)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314945
  • Cas Number : 338414-71-4
  • Molecular Weight : 307.309

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-310S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart