KEY465195052 Key Organics / BIONET

(3E)-3-[(2,4-difluorophenyl)imino]-1-(prop-2-yn-1-yl)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD02083094
  • Cas Number : 478031-70-8
  • Molecular Weight : 296.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71881

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-313S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart