KEY465195062 Key Organics / BIONET

ethyl 4-(4-fluorophenyl)-5-oxo-2-(trifluoromethyl)-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate

  • MDL number MFCD00974777
  • Cas Number : 338414-98-5
  • Molecular Weight : 383.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71969

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart