KEY465195064 Key Organics / BIONET

4-methyl-N'-[(4Z)-4,5,6,7-tetrahydro-1-benzofuran-4-ylidene]benzene-1-sulfonohydrazide

  • MDL number MFCD00794546
  • Cas Number : 338415-00-2
  • Molecular Weight : 304.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71979

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart